siRNA Details
| Gene Symbol: IRGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145805.1 | NP_001139277.1 | 2 | 1195 | ||
| XM_011537641.2 | XP_011535943.1 | 2 | 1195 | |||
| GenBank mRNA | BC128167.1 | - | 1 | 75 | 40113046 | |
| BC128168.1 | - | 1 | 82 | 40113047 | ||
| EU742619.1 | - | 2 | 1195 | |||

IRGM
Genomic Map