siRNA Details
| Gene Symbol: IQCA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304419.1 | NP_001291348.1 | 3, 4 | 501 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IQCA1L| Gene Symbol: IQCA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304419.1 | NP_001291348.1 | 3, 4 | 501 | ||