siRNA Details
Gene Symbol: KLRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198508.2 | NP_940910.1 | 5 | 1196 | ||
XM_005250311.3 | XP_005250368.1 | 4 | 1097 | |||
GenBank mRNA | AK126174.1 | - | 5 | 1201 | ||
BC104963.1 | - | 5 | 1161 | 8143966 | ||
BC104965.1 | - | 5 | 1161 | 8143968 | ||
BC110858.1 | - | 2 | 796 | 6139360 |
