siRNA Details
| Gene Symbol: KLRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198508.2 | NP_940910.1 | 1 | 446 | ||
| XM_005250311.3 | XP_005250368.1 | 1 | 451 | |||
| XM_011516140.2 | XP_011514442.1 | 1 | 451 | |||
| XM_011516141.2 | XP_011514443.1 | 1 | 451 | |||
| GenBank mRNA | AK126174.1 | - | 1 | 451 | ||
| BC104963.1 | - | 1 | 411 | 8143966 | ||
| BC104965.1 | - | 1 | 411 | 8143968 | ||
| BC110858.1 | - | 1 | 398 | 6139360 | ||
| DB128604.1 | - | 1 | 446 | |||

KLRG2
Genomic Map