siRNA Details
| Gene Symbol: CCNYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152523.2 | NP_689736.1 | 6, 7 | 582 | ||
| XM_005246343.1 | XP_005246400.1 | 5, 6 | 600 | |||
| GenBank mRNA | AK095479.1 | - | 1 | 21 | ||
| AK097751.1 | - | 6, 7 | 582 | |||

CCNYL1
Genomic Map