siRNA Details
| Gene Symbol: SUPT20HL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136233.1 | NP_001129705.1 | 1 | 2143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUPT20HL2| Gene Symbol: SUPT20HL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136233.1 | NP_001129705.1 | 1 | 2143 | ||