siRNA Details
| Gene Symbol: CXorf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013403.2 | NP_001013421.1 | 3 | 817 | ||
| GenBank mRNA | BC137148.1 | - | 3 | 817 | 9021145 | |
| BC137149.1 | - | 3 | 817 | 9021146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXorf66| Gene Symbol: CXorf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013403.2 | NP_001013421.1 | 3 | 817 | ||
| GenBank mRNA | BC137148.1 | - | 3 | 817 | 9021145 | |
| BC137149.1 | - | 3 | 817 | 9021146 | ||