siRNA Details
| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011719.1 | NP_001011719.1 | 5 | 853 | ||
| GenBank mRNA | AY875940.1 | - | 5 | 853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARSH| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011719.1 | NP_001011719.1 | 5 | 853 | ||
| GenBank mRNA | AY875940.1 | - | 5 | 853 | ||