siRNA Details
| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145020.1 | NP_001138492.1 | 3 | 239 | ||
| NR_026736.1 | 2 | 139 | ||||
| NR_026737.1 | 2 | 157 | ||||
| XM_006715088.3 | XP_006715151.1 | 3 | 408 | |||
| XM_011514572.2 | XP_011512874.1 | 2 | 204 | |||
| XM_011514574.2 | XP_011512876.1 | 2 | 204 | |||
| XM_011514575.2 | XP_011512877.1 | 3 | 390 | |||
| GenBank mRNA | AI809192.1 | - | 3 | 377 | ||
| AW293245.1 | - | 3 | 393 | |||
| BX280692.1 | - | 2 | 139 | |||

C6orf52
Genomic Map