siRNA Details
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318818.1 | NP_001305747.1 | 3, 4 | 306 | ||
NM_001318821.1 | NP_001305750.1 | 4, 5 | 383 | |||
NM_001318823.1 | NP_001305752.1 | 3, 4 | 306 | |||
NM_182612.3 | NP_872418.1 | 4, 5 | 383 | |||
NR_134867.1 | 3, 4 | 306 | ||||
XM_005252898.2 | XP_005252955.1 | 4, 5 | 368 | |||
XM_011520064.1 | XP_011518366.1 | 4, 5 | 365 | |||
XM_017017660.1 | XP_016873149.1 | 4, 5 | 368 | |||
XM_017017661.1 | XP_016873150.1 | 4, 5 | 365 | |||
XM_017017662.1 | XP_016873151.1 | 4, 5 | 365 | |||
XM_017017664.1 | XP_016873153.1 | 4, 5 | 368 | |||
XM_017017665.1 | XP_016873154.1 | 4, 5 | 368 | |||
XM_017017666.1 | XP_016873155.1 | 3, 4 | 289 | |||
XM_017017668.1 | XP_016873157.1 | 3, 4 | 289 | |||
GenBank mRNA | AK091602.1 | - | 4, 5 | 365 | ||
AK293559.1 | - | 3, 4 | 291 | |||
AK309153.1 | - | 3, 4 | 289 | |||
BC110868.1 | - | 4, 5 | 352 | 5726838 | ||
BC112145.1 | - | 4, 5 | 364 | 8327613 | ||
BC143418.1 | - | 4, 5 | 364 | 9051926 | ||
BC143419.1 | - | 3, 4 | 287 | |||
BC143422.1 | - | 3, 4 | 287 | |||
BC143423.1 | - | 3, 4 | 287 | |||
DA025348.1 | - | 4, 5 | 368 | |||
DC364167.1 | - | 4, 5 | 383 |
