siRNA Details
| Gene Symbol: GUSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027026.1 | 3 | 233 | |||
| NR_027027.1 | 3 | 233 | ||||
| NR_027028.2 | 3 | 233 | ||||
| GenBank mRNA | AK095656.1 | - | 3 | 157 | ||
| AK290693.1 | - | 3 | 200 | |||
| BC094818.1 | - | 3 | 172 | |||
| BC147000.1 | - | 3 | 187 | |||
| DA358891.1 | - | 3 | 233 | |||

GUSBP1
Genomic Map