siRNA Details
| Gene Symbol: DPY19L2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003561.2 | 19, 20 | 3192 | |||
| GenBank mRNA | AK131353.1 | - | 19, 20 | 3185 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPY19L2P2| Gene Symbol: DPY19L2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003561.2 | 19, 20 | 3192 | |||
| GenBank mRNA | AK131353.1 | - | 19, 20 | 3185 | ||