siRNA Details
| Gene Symbol: RTN4RL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178570.2 | NP_848665.1 | 3 | 1377 | ||
| GenBank mRNA | AF532858.1 | - | 3 | 1048 | ||
| AX751514.1 | - | 3 | 1048 | |||
| AY250221.1 | - | 3 | 1048 | |||
| BC113673.1 | - | 3 | 1070 | 8322725 | ||
| BC117276.1 | - | 3 | 1070 | 40125827 | ||
| BC143818.1 | - | 3 | 1070 | 9052333 | ||
| BE410139.1 | - | 1 | 53 | |||

RTN4RL2
Genomic Map