siRNA Details
| Gene Symbol: HSD11B1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267868.1 | NP_001254797.1 | 9 | 1627 | ||
| NM_001267869.1 | NP_001254798.1 | 6 | 1314 | |||
| NM_001267870.1 | NP_001254799.1 | 5 | 1154 | |||
| NM_001267871.1 | NP_001254800.1 | 6 | 1083 | |||
| NM_198533.2 | NP_940935.1 | 8 | 1326 | |||
| NM_198704.2 | NP_941993.1 | 6 | 1151 | |||
| NM_198705.2 | NP_941994.1 | 6 | 1246 | |||
| NM_198706.2 | NP_941995.1 | 8 | 1489 | |||
| NM_198707.2 | NP_941996.1 | 7 | 1426 | |||
| NM_198708.2 | NP_941997.1 | 7 | 1377 | |||
| GenBank mRNA | AK308206.1 | - | 9 | 1368 | ||
| AY268351.1 | - | 6 | 987 | |||
| AY268352.1 | - | 8 | 1190 | |||
| AY268353.1 | - | 7 | 1150 | |||
| AY268354.1 | - | 7 | 1078 | |||
| AY313894.1 | - | 8 | 1055 | |||
| AY313895.1 | - | 6 | 892 | |||
| AY313896.1 | - | 6 | 824 | |||
| BC053546.1 | - | 7 | 1007 | |||
| BC072434.1 | - | 6 | 1063 | |||
| BC094798.1 | - | 6 | 1070 | |||

HSD11B1L
Genomic Map