siRNA Details
| Gene Symbol: LOC374890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135233.1 | 2 | 266 | |||
| GenBank mRNA | BC030765.2 | - | 2 | 266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC374890| Gene Symbol: LOC374890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135233.1 | 2 | 266 | |||
| GenBank mRNA | BC030765.2 | - | 2 | 266 | ||