siRNA Details
| Gene Symbol: RBM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080504.2 | NP_001073973.2 | 8, 9 | 2338 | ||
| XM_005246075.1 | XP_005246132.1 | 8, 9 | 2679 | |||
| XM_011511152.2 | XP_011509454.1 | 8, 9 | 2679 | |||
| XM_011511153.2 | XP_011509455.1 | 8, 9 | 2679 | |||
| XM_017004054.1 | XP_016859543.1 | 8, 9 | 2679 | |||
| XM_017004055.1 | XP_016859544.1 | 8, 9 | 3049 | |||
| XM_017004056.1 | XP_016859545.1 | 8, 9 | 2619 | |||
| XM_017004057.1 | XP_016859546.1 | 8, 9 | 2679 | |||
| XM_017004058.1 | XP_016859547.1 | 3, 4 | 1017 | |||
| GenBank mRNA | BC126111.1 | - | 5, 6 | 421 | ||
| BX648107.1 | - | 7, 8 | 2991 | |||

RBM44
Genomic Map