siRNA Details
| Gene Symbol: COL18A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027498.1 | 3 | 511 | |||
| NR_028082.1 | 3 | 1595 | ||||
| GenBank mRNA | AK096071.1 | - | 3 | 511 | ||
| BC111539.2 | - | 3 | 1595 | 40026216 | ||
| BC111540.2 | - | 3 | 481 | 40026217 | ||

COL18A1-AS1
Genomic Map