siRNA Details
| Gene Symbol: IL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014336.1 | NP_001014358.1 | 3 | 266 | ||
| GenBank mRNA | AY499343.1 | - | 3 | 266 | ||
| BC132998.1 | - | 3 | 260 | 40147020 | ||
| BC133000.1 | - | 3 | 260 | 40147022 | ||

IL31
Genomic Map