siRNA Details
| Gene Symbol: C6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029863.2 | NP_001025034.1 | 1 | 513 | ||
| NM_001317342.1 | NP_001304271.1 | 2 | 273 | |||
| GenBank mRNA | AF451991.1 | - | 1 | 464 | ||
| AK295061.1 | - | 1 | 499 | |||
| AK295962.1 | - | 2 | 248 | |||
| BC051700.1 | - | 1 | 464 | 4792634 | ||

C6orf120
Genomic Map