siRNA Details
| Gene Symbol: TPTEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001591.1 | 8 | 1051 | |||
| GenBank mRNA | AY219889.1 | - | 8 | 1051 | ||
| BC107743.1 | - | 6 | 593 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTEP1| Gene Symbol: TPTEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001591.1 | 8 | 1051 | |||
| GenBank mRNA | AY219889.1 | - | 8 | 1051 | ||
| BC107743.1 | - | 6 | 593 | |||