siRNA Details
| Gene Symbol: TMEM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123376.2 | NP_001116848.1 | 1 | 216 | ||
| XM_011540071.2 | XP_011538373.1 | 1 | 225 | |||
| GenBank mRNA | AB235418.1 | - | 1 | 171 | ||
| BC127789.1 | - | 1 | 216 | 40133552 | ||
| BC127790.1 | - | 1 | 216 | 40133554 | ||
| BX538120.1 | - | 351 | ||||

TMEM72
Genomic Map