siRNA Details
| Gene Symbol: TMEM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123376.2 | NP_001116848.1 | 1, 2 | 259 | ||
| GenBank mRNA | AB235418.1 | - | 1, 2 | 214 | ||
| BC127790.1 | - | 1, 2 | 259 | 40133554 | ||
| BX538120.1 | - | 394 | ||||

TMEM72
Genomic Map