siRNA Details
| Gene Symbol: C10orf99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207373.2 | NP_997256.1 | 3 | 586 | ||
| XM_005269811.3 | XP_005269868.1 | 3 | 450 | |||
| GenBank mRNA | AI831407.1 | - | 1 | 246 | ||
| AY358751.1 | - | 3 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf99| Gene Symbol: C10orf99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207373.2 | NP_997256.1 | 3 | 586 | ||
| XM_005269811.3 | XP_005269868.1 | 3 | 450 | |||
| GenBank mRNA | AI831407.1 | - | 1 | 246 | ||
| AY358751.1 | - | 3 | 573 | |||