siRNA Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213606.3 | NP_998771.3 | 7 | 1436 | ||
| XM_017016237.1 | XP_016871726.1 | 6 | 1526 | |||
| XM_017016238.1 | XP_016871727.1 | 6 | 1465 | |||
| XM_017016239.1 | XP_016871728.1 | 6 | 1324 | |||
| GenBank mRNA | AK124901.1 | - | 3 | 715 | ||
| AK127303.1 | - | 7 | 1437 | |||
| BC086873.1 | - | 2 | 389 | |||
| CR936657.1 | - | 5 | 3634 | |||

SLC16A12
Genomic Map