siRNA Details
| Gene Symbol: FAM99A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026643.1 | 3 | 687 | |||
| GenBank mRNA | AK124823.1 | - | 1 | 1291 | ||
| CR627417.1 | - | 3 | 714 | |||
| Gene Symbol: FAM99B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026642.1 | 3 | 713 | |||

FAM99A
Genomic Map