siRNA Details
| Gene Symbol: SLC22A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039752.3 | NP_001034841.3 | 1 | 314 | ||
| XM_011545016.2 | XP_011543318.1 | 3 | 1319 | |||
| XM_017017700.1 | XP_016873189.1 | 3 | 876 | |||
| XM_017017701.1 | XP_016873190.1 | 3 | 1319 | |||
| XM_017017702.1 | XP_016873191.1 | 3 | 1319 | |||
| XM_017017704.1 | XP_016873193.1 | 3 | 875 | |||
| GenBank mRNA | BX091233.1 | - | 1 | 314 | ||

SLC22A10
Genomic Map