siRNA Details
| Gene Symbol: SPDYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008778.1 | NP_001008778.1 | 3, 4 | 246 | ||
| XM_017017705.1 | XP_016873194.1 | 2, 3 | 640 | |||
| XM_017017706.1 | XP_016873195.1 | 2, 3 | 640 | |||
| XM_017017707.1 | XP_016873196.1 | 2, 3 | 640 | |||
| XM_017017708.1 | XP_016873197.1 | 2, 3 | 640 | |||
| XM_017017709.1 | XP_016873198.1 | 2, 3 | 640 | |||
| XM_017017710.1 | XP_016873199.1 | 2, 3 | 640 | |||
| GenBank mRNA | AY820305.1 | - | 3, 4 | 246 | ||
| BC137244.1 | - | 3, 4 | 328 | 9021241 | ||
| BC137246.1 | - | 3, 4 | 328 | 9021243 | ||
| BC144548.1 | - | 3, 4 | 328 | 9053075 | ||

SPDYC
Genomic Map