siRNA Details
| Gene Symbol: LOC100506127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282456.2 | NP_001269385.1 | 1 | 824 | ||
| NM_001305225.1 | NP_001292154.1 | 1 | 824 | |||
| GenBank mRNA | AK095089.1 | - | 1 | 669 | ||
| BC040665.1 | - | 1 | 813 | |||

LOC100506127
Genomic Map