siRNA Details
| Gene Symbol: CCDC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025004.2 | NP_079280.2 | 15 | 2919 | ||
| GenBank mRNA | BU569785.1 | - | 4 | 481 | ||
| DB516555.2 | - | 4 | 372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC15| Gene Symbol: CCDC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025004.2 | NP_079280.2 | 15 | 2919 | ||
| GenBank mRNA | BU569785.1 | - | 4 | 481 | ||
| DB516555.2 | - | 4 | 372 | |||