siRNA Details
| Gene Symbol: SENCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038908.1 | 3 | 957 | |||
| GenBank mRNA | AI860159.1 | - | 1 | 359 | ||
| BC039676.1 | - | 3 | 957 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SENCR| Gene Symbol: SENCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038908.1 | 3 | 957 | |||
| GenBank mRNA | AI860159.1 | - | 1 | 359 | ||
| BC039676.1 | - | 3 | 957 | |||