siRNA Details
| Gene Symbol: LOC105369747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135024.1 | 1 | 49 | |||
| GenBank mRNA | BX114125.1 | - | 1 | 49 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369747| Gene Symbol: LOC105369747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135024.1 | 1 | 49 | |||
| GenBank mRNA | BX114125.1 | - | 1 | 49 | ||