siRNA Details
| Gene Symbol: PTPRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017019273.1 | XP_016874762.1 | 5 | 717 | ||
| XM_017019274.1 | XP_016874763.1 | 5 | 718 | |||
| XM_017019275.1 | XP_016874764.1 | 5 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTPRQ| Gene Symbol: PTPRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017019273.1 | XP_016874762.1 | 5 | 717 | ||
| XM_017019274.1 | XP_016874763.1 | 5 | 718 | |||
| XM_017019275.1 | XP_016874764.1 | 5 | 718 | |||