siRNA Details
| Gene Symbol: LOC101928617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135036.1 | 1, 2 | 161 | |||
| NR_135037.1 | 1, 2 | 161 | ||||
| GenBank mRNA | AL540933.3 | - | 1, 2 | 64 | ||
| BC044741.1 | - | 1, 2 | 161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928617| Gene Symbol: LOC101928617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135036.1 | 1, 2 | 161 | |||
| NR_135037.1 | 1, 2 | 161 | ||||
| GenBank mRNA | AL540933.3 | - | 1, 2 | 64 | ||
| BC044741.1 | - | 1, 2 | 161 | |||