siRNA Details
Gene Symbol: LOC101928617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135036.1 | 2 | 195 | |||
NR_135037.1 | 2 | 195 | ||||
GenBank mRNA | AL540933.3 | - | 2 | 98 | ||
BC044741.1 | - | 2 | 195 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135036.1 | 2 | 195 | |||
NR_135037.1 | 2 | 195 | ||||
GenBank mRNA | AL540933.3 | - | 2 | 98 | ||
BC044741.1 | - | 2 | 195 |