siRNA Details
| Gene Symbol: LINC01486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120462.1 | 2 | 316 | |||
| GenBank mRNA | AI150307.1 | - | 2 | 295 | ||
| BX107914.1 | - | 2 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01486| Gene Symbol: LINC01486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120462.1 | 2 | 316 | |||
| GenBank mRNA | AI150307.1 | - | 2 | 295 | ||
| BX107914.1 | - | 2 | 316 | |||