siRNA Details
| Gene Symbol: TMEM233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136534.1 | NP_001130006.1 | 1 | 445 | ||
| XM_005253880.3 | XP_005253937.1 | 1 | 1781 | |||
| XM_005253881.3 | XP_005253938.1 | 1 | 1777 | |||
| XM_011538328.2 | XP_011536630.1 | 1 | 1778 | |||
| XM_011538329.2 | XP_011536631.1 | 1 | 1778 | |||
| XM_011538330.2 | XP_011536632.1 | 1 | 1778 | |||
| XM_011538331.2 | XP_011536633.1 | 1 | 1776 | |||
| GenBank mRNA | AK296289.1 | - | 1 | 370 | ||

TMEM233
Genomic Map