siRNA Details
| Gene Symbol: MIPEPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038939.1 | 2 | 279 | |||
| NR_046461.1 | 2 | 279 | ||||
| GenBank mRNA | AK093279.1 | - | 2 | 231 | ||
| BG940872.1 | - | 2 | 99 | |||
| DA538853.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIPEPP3| Gene Symbol: MIPEPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038939.1 | 2 | 279 | |||
| NR_046461.1 | 2 | 279 | ||||
| GenBank mRNA | AK093279.1 | - | 2 | 231 | ||
| BG940872.1 | - | 2 | 99 | |||
| DA538853.1 | - | 2 | 280 | |||