siRNA Details
| Gene Symbol: LOC101927375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011535334.2 | XP_011533636.1 | 4, 5 | 692 | ||
| XM_011535335.2 | XP_011533637.1 | 4, 5 | 692 | |||
| XM_017020894.1 | XP_016876383.1 | 5, 6 | 769 | |||

LOC101927375
Genomic Map