siRNA Details
| Gene Symbol: SERP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010897.1 | NP_001010897.1 | 1, 2 | 275 | ||
| GenBank mRNA | BC029067.1 | - | 1, 2 | 275 | 5197847 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERP2| Gene Symbol: SERP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010897.1 | NP_001010897.1 | 1, 2 | 275 | ||
| GenBank mRNA | BC029067.1 | - | 1, 2 | 275 | 5197847 | |