siRNA Details
| Gene Symbol: LINC00433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047021.1 | 1 | 452 | |||
| GenBank mRNA | BI520070.1 | - | 1 | 458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00433| Gene Symbol: LINC00433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047021.1 | 1 | 452 | |||
| GenBank mRNA | BI520070.1 | - | 1 | 458 | ||