siRNA Details
| Gene Symbol: NOVA1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI278049.1 | - | 1, 2 | 233 | ||
| N48783.1 | - | 1, 2 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOVA1-AS1| Gene Symbol: NOVA1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI278049.1 | - | 1, 2 | 233 | ||
| N48783.1 | - | 1, 2 | 240 | |||