siRNA Details
| Gene Symbol: LOC100506700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109999.1 | 1 | 239 | |||
| GenBank mRNA | AI078294.1 | - | 3 | 414 | ||
| CR743158.1 | - | 1 | 239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506700| Gene Symbol: LOC100506700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109999.1 | 1 | 239 | |||
| GenBank mRNA | AI078294.1 | - | 3 | 414 | ||
| CR743158.1 | - | 1 | 239 | |||