siRNA Details
| Gene Symbol: C15orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207380.2 | NP_997263.2 | 11 | 1262 | ||
| GenBank mRNA | AK125329.1 | - | 11 | 1255 | ||
| AK126035.1 | - | 7 | 652 | |||
| BC140897.1 | - | 7 | 651 | 9021765 | ||
| BC140902.1 | - | 7 | 651 | 9021770 | ||
| CR749408.1 | - | 8 | 956 | |||

C15orf52
Genomic Map