siRNA Details
| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 1 | 602 | |||
| NR_028140.1 | 1 | 117 | ||||
| GenBank mRNA | AK124283.1 | - | 1 | 597 | ||
| BC028144.1 | - | 1 | 113 | 5214282 | ||
| BC132704.1 | - | 1 | 60 | |||
| BC132706.1 | - | 1 | 60 | |||

SPATA41
Genomic Map