siRNA Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242757.1 | NP_001229686.1 | 7 | 1219 | ||
| XM_011522897.2 | XP_011521199.1 | 8 | 1267 | |||
| XM_011522899.2 | XP_011521201.1 | 8 | 1204 | |||
| XM_017022962.1 | XP_016878451.1 | 5 | 869 | |||
| XM_017022963.1 | XP_016878452.1 | 5 | 2617 | |||
| XM_017022964.1 | XP_016878453.1 | 6 | 1773 | |||
| XM_017022965.1 | XP_016878454.1 | 5 | 2558 | |||
| XM_017022966.1 | XP_016878455.1 | 6 | 2396 | |||
| GenBank mRNA | AL137382.1 | - | 2 | 72 | ||

SLC22A31
Genomic Map