siRNA Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242757.1 | NP_001229686.1 | 2, 3 | 560 | ||
| XM_011522897.2 | XP_011521199.1 | 3, 4 | 608 | |||
| XM_011522899.2 | XP_011521201.1 | 3, 4 | 545 | |||
| XM_017022966.1 | XP_016878455.1 | 2, 3 | 1512 | |||

SLC22A31
Genomic Map