siRNA Details
| Gene Symbol: C17orf98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080465.2 | NP_001073934.1 | 2 | 237 | ||
| GenBank mRNA | DY654789.1 | - | 2 | 188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17orf98| Gene Symbol: C17orf98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080465.2 | NP_001073934.1 | 2 | 237 | ||
| GenBank mRNA | DY654789.1 | - | 2 | 188 | ||