siRNA Details
Gene Symbol: RPRML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203400.4 | NP_981945.1 | 1 | 480 | ||
GenBank mRNA | BC033942.1 | - | 1 | 485 | 5274621 | |
BC108690.1 | - | 1 | 455 | 6140781 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RPRML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203400.4 | NP_981945.1 | 1 | 480 | ||
GenBank mRNA | BC033942.1 | - | 1 | 485 | 5274621 | |
BC108690.1 | - | 1 | 455 | 6140781 |