siRNA Details
| Gene Symbol: LINC01483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109971.1 | 3 | 301 | |||
| NR_109972.1 | 3 | 301 | ||||
| GenBank mRNA | BC068583.1 | - | 3 | 242 | ||
| BC094791.1 | - | 3 | 242 | |||
| DA832262.1 | - | 3 | 301 | |||
| DA856452.1 | - | 3 | 301 | |||

LINC01483
Genomic Map