siRNA Details
| Gene Symbol: LOC101928674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110847.1 | 1, 2 | 328 | |||
| GenBank mRNA | BG437436.1 | - | 1, 2 | 324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928674| Gene Symbol: LOC101928674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110847.1 | 1, 2 | 328 | |||
| GenBank mRNA | BG437436.1 | - | 1, 2 | 324 | ||